微孔板离心机,PCR Plate Spinner
供应商: VWR BRAND PRIVATE LABEL USA
89184-610
VWRU89184-610EA 21880 CNY
VWRU89184-610
微孔板离心机,PCR Plate Spinner
离心机 台式离心机
The PCR plate spinner offers a personal-sized centrifuge designed to spin down PCR plates.
  • Space saving design; quiet, simple operation
  • Easy plate loading system
  • Quickly spins down droplets and condensation
  • Use before and after thermal cycling to improve PCR yield
  • Cold room compatible
This unique centrifuge is equipped with a dual-position vertical rotor that accepts a wide variety of PCR plates; 96-well, 384-well, skirted and non skirted. The rotor features integral plate guides, ensuring that the plates stay vertical and do not tip inside the rotor.
现在订购
规格
型号 PCR Plate Spinner
Environmentally Preferable
最大容量 2 PCR plates
最大速度 2500 min⁻¹
最大相对离心力 500 ×g
电力供应 230 V / 50 - 60 Hz
W×D×H 191×208×183 mm
Weight 3 kg