VWR® Labmax, Bottle-Top Dispensers
供应商: VWR BRAND PRIVATE LABEL USA
82017-770 40000-060
VWRU82017-770EA 16035 CNY
VWRU82017-770 VWRU40000-060
VWR® Labmax, Bottle-Top Dispensers
瓶口分配器
Variable volume bottle-top dispenser with locking rocker switch, autoclavable.
  • Unique closed circuit air purging system for air and bubbles elimination
  • Exclusive integrated swivel neck
  • Reagent 'guard valve' for safety shut down
  • Patented direct displacement PTFE piston
  • Auto-calibrating, quick and accurate volume adjustment
The dispensers can be disassembled quickly and easily for thorough cleaning. Quick and accurate volume adjustment is easy with the locking rocker switch. The zero point is manufactured into the Labmax™; calibration is not necessary,More Product Information