VWR® Advanced Magnetic Hotplate Stirrers, 230 V
供应商: VWR BRAND PRIVATE LABEL USA
76447-018 76447-038 76447-054 76447-030
VWRU76447-018EA 12556 CNY
VWRU76447-018 VWRU76447-038 VWRU76447-054 VWRU76447-030
VWR® Advanced Magnetic Hotplate Stirrers, 230 V
搅拌器 加热磁力搅拌器
Advanced magnetic hotplate stirrers are designed to deliver accurate and repeatable results in all research, academic, and industrial applications. With temperature ranges up to 500 °C and stirring speeds reaching 1600 min⁻¹, the VWR® hotplate stirrers provides proper mixing and superior temperature control. Available in three sizes and a choice of two top plate materials.
  • 230 V model
  • Excellent temperature uniformity and powerful stirring
  • Easy to read, large LCD displays temperatures and speeds
  • Heating models include probe connection for optional
More Product Information