VWR® Professional Hotplate-Stirrers
供应商: VWR BRAND PRIVATE LABEL USA
76447-020 76447-022 76447-042 76447-046
VWRU76447-020EA 23061 CNY
VWRU76447-020 VWRU76447-022 VWRU76447-042 VWRU76447-046
VWR® Professional Hotplate-Stirrers
搅拌器 加热磁力搅拌器
Designed for applications requiring exceptional accuracy, stability, and repeatability, these hotplate-stirrers are equipped with superior heating and mixing capabilities. With temperature ranges up to 500 °C and stirring speeds reaching 1600 rpm, the VWR® hotplate-stirrers are equipped to handle any research, academic and industrial application. Available in two sizes.
  • Excellent temperature uniformity and powerful stirring
  • Easy to read, large LCD displays temperatures, speed and timer settings
  • Cool touch, chemical-resistant housing
  • Includes an RTD
More Product Information