VWR®, Mini Centrifuges
供应商: VWR BRAND PRIVATE LABEL USA
76269-066
VWRU76269-066EA 5025 CNY
VWRU76269-066
VWR®, Mini Centrifuges
离心机 微型离心机
The VWR® Mini centrifuge is designed for quick spin-downs of microcentrifuge tubes and PCR strips and tubes.
  • Quick-release rotors; no tools required
  • Eight position microtube rotors
  • Spins up to 7500 rpm/3200 ×g
  • Decelerates in less than one second
  • Two-year warranty
The VWR® Mini Centrifuge features two quick-release rotors (no tools required): a 8×1,5/2,0 ml rotor and a 32×0,2 ml PCR rotor which also accommodates 4 eight place PCR strips. Close the lid and the unit quickly accelerates to 7500 rpm (3200 rcf). Open the lid and the rotor quickly andMore Product Information
现在订购
规格
Environmentally Preferable
容量 8×1,5/2,0 + 32×0,2 ml
尺寸 178×152×127 mm