VWR® Traceable®, Digital thermometers, long-stem
供应商: VWR BRAND PRIVATE LABEL USA
61220-416
VWRU61220-416EA 910 CNY
VWRU61220-416
VWR® Traceable®, Digital thermometers, long-stem
温度计 数字温度计
Constructed of ABS plastic and stainless-steel, these thermometers are ideal for replacing all glass and mercury thermometers in the lab. Perfect-sized unit fits into flasks, cuvettes, test tubes, beakers, ovens, and incubators. Read temperatures in foods, gases, samples, reagents, water baths and semisolids.
  • On/off switch provides longer battery life, °F/°C switchable range
  • Long stem allows for taking readings while stirring
  • Readings are updated every second
With stainless steel probe.
Certifications: Compliance with ISO/IEC 17025; accredited by A2More Product Information