VWR®Traceable®, Digital timer, two channels, jumbo
供应商: VWR BRAND PRIVATE LABEL USA
61161-340
VWRU61161-340EA 534 CNY
VWRU61161-340
VWR®Traceable®, Digital timer, two channels, jumbo
定时器 数字计时器
This timer is ideal for catching test results, signaling time intervals, and alerting appointments.
  • Timing capacity: Count up or down from 23 hours, 59 minutes, 59 seconds
  • Simultaneously maintains two separate countdown timing channels with distinct alarms, stopwatch, and time-of-day options
  • Channels may be programmed individually or simultaneously
  • Alarm timer features large, 2,5 cm (1") digits for 'across the room' readability
  • Alarm may be turned off manually or silences automatically after one minute
  • Just touch a button to return the display to the last
More Product Information