VWR®, Petri Dish Incubation Trays, Stackable
供应商: VWR BRAND PRIVATE LABEL USA
60914-210
VWRU60914-210EA 1410 CNY
VWRU60914-210
VWR®, Petri Dish Incubation Trays, Stackable
架子 培养箱架子
Stackable Petri dish incubation trays increase overall capacity of an incubator by holding five 100 mm diameter or fifteen 60mm diameter Petri dishes.
  • Holes in tray bottom facilitate air flow
  • Polypropylene construction can be disinfected with 70% ethanol or common laboratory disinfectants
Each tray includes two large label areas for critical sample identification by marker or label.
Caution: Not recommended for use above 100 °C. Autoclaving may warp.