洁净室实验服,VWR® Maximum,SF
供应商: VWR Collection
414004-703
VWRI113-1143EA 1045 CNY
VWRI113-1143 VWRI113-1142 VWRI113-1141 VWRI113-1140 VWRI113-1139 VWRI113-1138 VWRU414004-703
洁净室实验服,VWR® Maximum,SF
Disposable Garments
VWR® Maximum SF lab coats are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness, and comfort properties.
  • Tapered collar, inset sleeves, snap closure, elasticated cuffs and three pockets
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding: Level I Helmke drum classification
  • Light chemical splash resistance
  • Improved breathability to ensure user
More Product Information