VWR® Maximum,SF洁净室防护罩袍
供应商: VWR Collection
414004-702
VWRI113-1172EA
1245
CNY
VWRI113-1172
VWRI113-1171
VWRI113-1170
VWRI113-1169
VWRI113-1167
VWRI113-1168
VWRU414004-702
VWR® Maximum,SF洁净室防护罩袍
Disposable Garments
规格
Environmentally Preferable | |
关闭类型 | 带按扣紧固 |
袋 | 无口袋 |
Gender | 中性 |
Garment application | Cleanroom,Laboratory |
Weight | 63 g/m² |