洁净室防护服,VWR® Maximum,SF
供应商: VWR Collection
414004-308 414004-683 414004-684 414004-685 414004-686
VWRI113-1076EA 1418 CNY
VWRI113-1076 VWRI113-1075 VWRI113-1074 VWRI113-1079 VWRI113-1086 VWRI113-1089 VWRI113-1077 VWRI113-1087 VWRI113-1078 VWRI113-1085 VWRI113-1084 VWRI113-1082 VWRI113-1081 VWRI113-1080 VWRI113-1088 VWRU414004-308 VWRU414004-683 VWRU414004-684 VWRU414004-685 VWRU414004-686 VWRI113-9161
洁净室防护服,VWR® Maximum,SF
Disposable Garments
VWR® Maximum SF overalls are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
  • Available as a standard overall with collar, either elasticated ankles or non slip boot covers, or hooded overall with fluid-impervious boot covers
  • Inset sleeves, zip closure, elasticated cuffs
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding:
More Product Information