VWR®, Hygiene Masks, Advanced Protection
供应商: VWR BRAND PRIVATE LABEL USA
414004-662 414004-666
VWRU414004-662EA 406.85 CNY
VWRU414004-662 VWRU414004-666
VWR®, Hygiene Masks, Advanced Protection
口罩 卫生口罩
VWR® Advanced Protection face masks feature a three-ply construction for excellent particle and bacterial filtration efficiency. Available in two styles, with spandex ear loops or PP ties ultrasonically welded to maintain softness and protect against particulates.
  • Latex-free
  • Guards against bacteria and particulates
  • Highly breathable and fluid-resistant
  • Malleable 12 cm noseband ensures custom, secure seal
  • Soft and hypoallergenic
Certifications: Masks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls toMore Product Information