洁净室鞋套,防滑,VWR 高级款,SF
供应商: VWR Collection
414004-498 414004-499 414004-500
VWRI113-1383EA 719.89 CNY
VWRI113-1383 VWRI113-1382 VWRI113-1378 VWRI113-1377 VWRI113-1384 VWRU414004-498 VWRU414004-499 VWRU414004-500
洁净室鞋套,防滑,VWR 高级款,SF
Disposable Garments
VWR® 高级款 SF鞋套采用纺粘丙纶织物(SPP)制造,带透气性聚乙烯薄膜涂层。
  • High coefficient of friction to provide excellent traction
  • Water-resistant
  • Durable fabric with good breathability
  • Significant fluid and particulate barrier
  • Particle shedding: Level II Helmke drum classification
  • Soft, cloth-like fabric
在经ISO认证的设施中,在严格的过程控制下进行测试和制造。通过独立的实验室测试进行批次控制和验证。
长度:394 mm (U*), 432 mm (XL), 470 mm (XXL).
*通用尺寸.
现在订购
规格
Environmentally Preferable
Gender
Garment application Cleanroom,Laboratory