VWR® Durac™, Baumé hydrometer
供应商: VWR BRAND PRIVATE LABEL USA
34670-161
VWRU34670-161EA 555 CNY
VWRU34670-161
VWR® Durac™, Baumé hydrometer
液体比重计
Baumé hydrometer designed for determining the specific gravity and Baumé in liquids lighter then water.
  • Dual scale
  • Individually serialised
  • Lead free metal ballast
Specially used in chemistry, pharmacology, food and beverage, and in the starch industry.
Certifications: NIST and DAkkS. Calibration conforms to A2LA accreditions.
Delivery information: Supplied with a multi-language statement of accuracy.
现在订购
规格
Environmentally Preferable
Reference temperature