VWR®, Orbital Mini Shakers
供应商: VWR BRAND PRIVATE LABEL USA
12620-938 10027-122
VWRU12620-938EA 19374 CNY
VWRU12620-938 VWRU10027-122
VWR®, Orbital Mini Shakers
振荡器 轨道振荡器
Variable-speed, microprocessor control provides consistent, uniform mixing action.
  • Digital display for speed and time
  • Microprocessor control
  • Timer with audible alarm
Ramping feature slowly increases speed to set point for added safety. Independent LED display features speed and time. Timer will display elapsed time, or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero. Ideal for use in incubators, CO2 incubators, or cold rooms. Durable base is constructed of cast aluminum forMore Product Information
现在订购
规格
型号 Mini shaker Mini shaker
Environmentally Preferable
Speed 100 - 1200 min⁻¹ 100 - 1200 min⁻¹
摇动 Orbital Orbital
Max. load 3,6 kg 3,6 kg
轨道 3 mm 3 mm
重量 11,3 kg 11,3 kg
W×D×H 279×432×102 mm 279×432×102 mm
Platform W×D 218×297 mm 218×297 mm
电气 120 V, 50/60 Hz 320 V, 50/60 Hz
计时器 1 second – 160 hours 1 second - 160 hours