VWR®, Laboratory Freezer, Chest Style, Manual Defrost Series
供应商: VWR BRAND PRIVATE LABEL USA
10819-422
VWRU10819-422EA 131232 CNY
VWRU10819-422
VWR®, Laboratory Freezer, Chest Style, Manual Defrost Series
冷柜 实验室通用冰箱
Freezer that features keyed door locks, probe access port, and sample-simulated probe.
  • Resettable one-point min/max history
  • Temperature controller
  • Digital setting accuracy
  • Precise temperature maintenance
  • Independent operation for security
Units are equipped with a sample-simulated temperature display, high/low temperature alarm, sensor error alarm, power failure alarm, alarm mute with ring back, remote alarm contacts and alarm validation.
Certifications: U/C-UL listed.
Delivery information: Units are supplied with a two-years parts and labourMore Product Information