VWR®, Wide Mouth Volumetric Flask with PE Stopper, Class A, Serialised
供应商: VWR BRAND PRIVATE LABEL USA
10124-112 10124-196
VWRU10124-112EA 6563 CNY
VWRU10124-112 VWRU10124-196
VWR®, Wide Mouth Volumetric Flask with PE Stopper, Class A, Serialised
烧瓶 容量瓶
Made from borosilicate 3.3 expansion glass.
  • Individually serialised
  • Wide-mouthed design
Flask is serialised and supplied with polyethylene stopper.
Delivery information: Flask comes with individual certificate of compliance.