VWR® BCR® Cleanroom Bond Paper
供应商: VWR USA
77788-016
VWRM77788-016EA 0 CNY
VWRM77788-016
VWR® BCR® Cleanroom Bond Paper
纸张
Designed to minimise contamination and run through copiers, laser printers, and impact printers.
  • Purest cellulose to minimise contamination
  • Specialized coating to reduce particles, fibers, and micro-organisms
  • Retains toner from copiers and laser printers to reduce smudging
  • Autoclavable
  • Latex-free
  • ISO class 3 to 8
  • Cleanroom class 1 to 100000
Optimal strength and durability ensures maximum consistency in printing and photocopying. A cellulose base sheet with a synthetic latex binder is added during manufacturing to reduce the potential of particle orMore Product Information