VWR®, Cleanroom Spiral Notebooks
供应商: VWR Collection
VWRI818-0279EA 3061 CNY
VWRI818-0279 VWRI818-0278 VWRI818-0280
VWR®, Cleanroom Spiral Notebooks
笔记本
These cleanroom spiral notebooks are made of synthetic latex paper.
  • White
  • 100 pages
  • Polypropylene cover
  • Spiral bound
  • 85 gsm
Suitable for cleanroom use.