VWR® Rigid PCR Plates
供应商: VWR Collection
VWRI732-4396EA 0 CNY
VWRI732-4396 VWRI732-4397 VWRI732-4398 VWRI732-4399 VWRI732-4400 VWRI732-4401 VWRI732-4402 VWRI732-4404 VWRI732-4406 VWRI732-4407 VWRI732-4408 VWRI732-4409 VWRI732-4410 VWRI732-4411 VWRI732-4412 VWRI732-4413 VWRI732-4414 VWRI732-4415 VWRI732-4416 VWRI732-4417 VWRI732-4418 VWRI732-4421 VWRI732-4423 VWRI732-4424
VWR® Rigid PCR Plates
微孔板 PCR板
VWR® Rigid PCR Plates feature a rugged polycarbonate frame, providing a solid and flat surface that guarantees seamless interaction with automated liquid handling platforms. With a firm grip assured, robots can confidently place VWR® Rigid PCR Plates with utmost precision, eliminating any concerns of misalignment.
  • Polycarbonate frame is rigid and flat to facilitate a firm grip by robotic arms and ensure reliable placement
  • Clear polypropylene wells provide optimal thermal transfer and higher reaction efficiency
  • White well options are available to enhance
More Product Information