VWR®, PCR/qPCR Plates, 96-Wells, Plastic
供应商: VWR Collection
VWRI732-3753EA 1366 CNY
VWRI732-3753 VWRI732-3754 VWRI732-3755 VWRI732-3756 VWRI732-3757 VWRI732-3758 VWRI732-3759 VWRI732-3760 VWRI732-3761 VWRI732-3762 VWRI732-3763 VWRI732-3764 VWRI732-3765 VWRI732-3766 VWRI732-3767 VWRI732-3768 VWRI732-3769 VWRI732-3770
VWR®, PCR/qPCR Plates, 96-Wells, Plastic
微孔板 PCR板
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results.
  • Natural or white PP
  • Standard or low profile
  • Flat top or raised deck
  • Non skirted, semi-skirted or fully skirted
  • Options to fit standard, ABI FAST and Roche 96-well blocks
  • Autoclavable
96-well PCR plates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.
Certifications: FDA 21 CFR compliance. Compliance to BSE/TSE (free from animal origin). Non pyrogenic, DNase-, RNase- and latex-free.
订购须知 Cap stripsMore Product Information