VWR®, Polystyrene Assay Plates, 96- and 384-Well
供应商: VWR Collection
VWRI732-3720EA 3142 CNY
VWRI732-3720 VWRI732-3721 VWRI732-3722 VWRI732-3723 VWRI732-3724 VWRI732-3725 VWRI732-3728 VWRI732-3729 VWRI732-3730 VWRI732-3731 VWRI732-3734 VWRI732-3735 VWRI732-3736 VWRI732-3737 VWRI732-3738 VWRI732-3739 VWRI732-3740 VWRI732-3741 VWRI732-3742 VWRI732-3743 VWRI732-3744 VWRI732-3745 VWRI732-3746 VWRI732-3747 VWRI732-3748 VWRI732-3749 VWRI732-3750
VWR®, Polystyrene Assay Plates, 96- and 384-Well
微孔板 高温超导微孔板
Manufactured from high quality crystal polystyrene. They are ideal for routine assays, mixing and storage.
  • Flat bottom ideal for microscopic applications
  • V-bottom minimising residual liquid
  • U-bottom for optimising sample/cell collection
VWR® assay plates are available in clear, white or black polystyrene, with solid or clear bottom.
The bottom of the clear bottom plates is just 0,4 mm thick and extremely flat (±0,1 mm tolerance), ensuring high measurement precision and elimination of reading errors when performing cell-based assays usingMore Product Information