VWR®, Combined pH Electrodes, pHASE
供应商: VWR Collection
VWRI662-1799EA 4848 CNY
VWRI662-1799 VWRI662-1800 VWRI662-1801 VWRI662-1778 VWRI662-1780 VWRI662-1781 VWRI662-1782
VWR®, Combined pH Electrodes, pHASE
电极 pH电极
Combined electrodes providing fast, accurate and stable pH measurements, ideal for control quality and research and development use.
  • Fast, accurate and stable pH measurements, fast thermal equilibration, high accuracy even in samples with large temperature range
  • Available with sealed design (economical, easy to use, with minimum maintenance) or Rapid-Renew design (refillable electrodes used with dirty or other samples that cause junction fouling)
  • Available with epoxy body (with good chemical resistance, maximum ruggedness and removable bulb guard) or glass
More Product Information