VWR®, Macro Cuvettes
供应商: VWR Collection
VWRI634-1067EA 278 CNY
VWRI634-1067
VWR®, Macro Cuvettes
比色皿 分光光度计比色皿
Disposable macro cuvettes, four clear sides.
  • Clear and clean optical path
  • Four windows for precise measurements
  • Manufactured under controlled room conditions
Specially designed for fluorescence spectroscopy.