带载玻片盒
供应商: VWR Collection
VWRI612-2706EA 140 CNY
VWRI612-2706 VWRI612-2707 VWRI612-2708 VWRI631-1517 VWRI631-1519 VWRI631-1521 VWRI631-1522 VWRI631-1524 VWRI631-1526 VWRI631-1506 VWRI631-1507 VWRI631-1508 VWRI631-1510 VWRI631-1500 VWRI631-1501 VWRI631-1502 VWRI631-1503 VWRI631-1504
带载玻片盒
显微镜幻灯片盒
Heavy-duty, two-piece microscope slide boxes are designed to store standard slides for transport.
  • Colour coded for easy identification
  • Slide inventory sheet on inside of the cover
  • Separate lid (box for 25 slides)
  • Lid with plastic snap closure (box for 50 slides)
  • Nickel plated clasp and hinge pin (box for 100 slides)
  • Each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box
  • Moulded, rigid box: Securely holds slides in grooved slots
  • Stackable for long-term storage
The box is made of durable ABS and secured with aMore Product Information