载玻片盒,Q Path®
供应商: VWR Collection
VWRI631-1079EA 2684 CNY
VWRI631-1079
载玻片盒,Q Path®
显微镜幻灯片盒
This box for five slides prevents items from breaking or sticking. Can be used for applications in cytology, such as conventional smears.
W×D×H: 82×30×18 mm