VWR®, IOS W-Plan Objectives
供应商: VWR Collection
VWRI630-2949EA 1089 CNY
VWRI630-2949 VWRI630-2948 VWRI630-2950 VWRI630-2954 VWRI630-2955 VWRI630-2951 VWRI630-2952 VWRI630-2953
VWR®, IOS W-Plan Objectives
显微镜物镜
These high contrast objectives are ideal for routine observations in laboratories.
  • Ensure field flatness up to F.N. 22
  • Based on infinity-corrected optical system
  • 100× is also available with iris for darkfield