VWR® Durac™ Plus, Hydrometers
供应商: VWR Collection
VWRI620-2388EA 3650 CNY
VWRI620-2388 VWRI620-2389
VWR® Durac™ Plus, Hydrometers
液体比重计
These plain form hydrometers are designed for applications where rough handling or contamination could be an issue.
  • Durable and shatterproof, lead free ballast
  • Easy to read black scale on yellow back to avoid eye fatigue
  • Individually serialised - Temperature standardisation of 15 °C
Suitable for applications in the food or beverage industries.
Certifications: NIST.
订购须知 Includes a multi-language statement of accuracy.
Caution: Not recommended for use with petroleum-based liquids or other corrosives, these should be stored in a clean, dry,More Product Information