分配器吸头,通用
供应商: VWR Collection
VWRI613-1032EA 0 CNY
VWRI613-1032 VWRI613-1033 VWRI613-5867 VWRI613-5868 VWRI613-5869 VWRI613-5870 VWRI613-5871 VWRI613-5872 VWRI613-5873 VWRI613-5874 VWRI613-5875 VWRI613-5877 VWRI613-5878 VWRI613-5879 VWRI613-5880 VWRI613-5881 VWRI613-5882 VWRI613-5883 VWRI613-5884
分配器吸头,通用
吸头
By using the direct displacement method, these tips work well for infectious and volatile liquids, as the fluid never will get in touch with the instrument which avoids any contamination.
  • Compatible with: VWR® Stepper and Stepper Pro, Ritter Ripette®, and Ripette® pro, Brand HandyStep®, HandyStep® electronic and HandyStep® touch*, Eppendorf® Multipette® 4780 and Multipette® plus, Gilson Repetman®, EasyStep, Minilab 100/101, STEPMATE Handrop (* without activated display/tip recognition)
  • Sustainable design based on virgin resins
  • Nine different tip sizes
More Product Information