VWR® Table Windshield for Balances
供应商: VWR Collection
VWRI611-5598EA 20047 CNY
VWRI611-5598
VWR® Table Windshield for Balances
天平
Protect laboratory balances with a removable windshield during measurements.
  • Large windshield with a white metal frame
  • Anti-static acrylic with two side openings
This windshield ensures accurate measurements while safeguarding against environmental changes for Analytical, Semimicro, and especially Microbalances.