VWR®, Platform Scales
供应商: VWR Collection
VWRI611-5085EA 21414 CNY
VWRI611-5085 VWRI611-5074 VWRI611-5069 VWRI611-5071 VWRI611-5075 VWRI611-5078 VWRI611-5079 VWRI611-5077 VWRI611-5080 VWRI611-5086 VWRI611-5081 VWRI611-5088
VWR®, Platform Scales
天平
Industrial series of advanced platform scales, offering increased accuracy and fast stabilisation time, combined with easy, intuitive operation and a large graphical screen for laboratory applications and industrial settings. Ideal for weighing, mixing, counting and check weighing.
  • Efficient filtering algorithms to deliver quick and accurate results
  • Fast stabilisation time
  • Easy to clean and high chemical resistance
  • Choice of two platform sizes (M-Line 280×280 mm, L-Line 300×400 mm)
  • Different column size, for tabletop or ground weighing (M-
More Product Information