VWR® SMG Advanced Semi-Micro Balances, Graphic Display
供应商: VWR Collection
VWRI611-4952EA 44219 CNY
VWRI611-4952 VWRI611-4953 VWRI611-4954 VWRI611-4955
VWR® SMG Advanced Semi-Micro Balances, Graphic Display
天平
Semi-micro balances with resolution of 0,01 mg, they are equipped with a new highly integrated electromagnetic cell. All models feature automatic internal calibration and are characterised by a high level of precision and repeatability. These VWR® semi-micro balances provide an excellent price/performance ratio, thanks to continuous intensive research and study of how weighing works. All models have a stainless steel top pan.
  • Integrated weighing sensor for enhanced stability and quick response time
  • Selectable filter levels and measuring units
  • Check
More Product Information