VWR® IS High Capacity Balances
供应商: VWR Collection
VWRI611-4530EA 16025 CNY
VWRI611-4530 VWRI611-4549 VWRI611-4550 VWRI611-4536 VWRI611-4528 VWRI611-4537 VWRI611-4529 VWRI611-4539 VWRI611-4557 VWRI611-4558 VWRI611-4559 VWRI611-4540 VWRI611-4538 VWRI611-4556 VWRI611-4547 VWRI611-4548 VWRI611-4520 VWRI611-4546 VWRI611-4946 VWRI611-4947 VWRI611-4948 VWRI611-4949 VWRI611-4950 VWRI611-4951
VWR® IS High Capacity Balances
天平
The IS range of high capacity balances are robust, reliable and ensure high performance. Models are equipped with either a backlit LCD with adjustable contrast, a large graphic display or a large 5" colour touch screen, the balances have been designed to deliver top performance at competitive prices.
  • Electromagnetic force restoration weighing system (for 0,01 g resolution)
  • High readability load cell weighing system (for 0,1 g resolution)
  • Solidly built with an ergonomic keypad
  • External calibration
  • Measuring units are selectable and customisable
  • Hook for
More Product Information