分析天平,LA系列
供应商: VWR Collection
VWRI611-2258EA 11567 CNY
VWRI611-2258 VWRI611-2260 VWRI611-2261 VWRI611-2263 VWRI611-2264 VWRI611-2265 VWRI611-2269 VWRI611-2271 VWRI611-3123 VWRI611-3261 VWRI611-3263 VWRI611-3264 VWRI611-3282 VWRI611-3280 VWRI611-3326 VWRI611-3328 VWRI611-3343 VWRI611-3350 VWRI611-3351 VWRI611-3449 VWRI611-3451 VWRI611-3546 VWRI611-4281 VWRI611-5615
分析天平,LA系列
天平
An economical range of reliable, high performance analytical balances. All models have a stainless steel top pan.
  • Check weighing (alarm function), weight totalising, density determinations of samples and piece counting functions
  • Option of external or internal calibration, RS232 included as standard
  • Weighing chamber L×W×H: 162×171×225 mm
User can choose three options for models with internal calibration (denoted by 'i' at end of the model name):
Balance will calibrate using the internal weight by pressing CAL button
Balance will automatically recalibrateMore Product Information