VWR®, Septa ND9
供应商: VWR Collection
VWRI548-3323AEA 889 CNY
VWRI548-3323A VWRI548-3324A VWRI548-3293A VWRI548-3325A VWRI548-3326A VWRI548-1564A VWRI548-0182A VWRI548-0183A VWRI548-2515A
VWR®, Septa ND9
隔垫
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.