试管架
供应商: VWR Collection
VWRI525-0192EA 1539 CNY
VWRI525-0192 VWRI525-0193 VWRI525-0194 VWRI525-0195
试管架
架子 Tube Racks
PTFE.
  • 优良的耐化学性
  • 温度耐受范围 −200 至+280℃
  • 非粘性表面,易于清洁
Robust and inert to virtually all chemical reagents, with very good thermal resistance. Suitable for use in cryogenics, freezers and ovens.