VWR®, Syringe Filters
供应商: VWR Collection
VWRI514-1251EA 193 CNY
VWRI514-1251 VWRI514-1249 VWRI514-1250 VWRI514-1240 VWRI514-1241 VWRI514-1243 VWRI514-1244 VWRI514-1245 VWRI514-1246 VWRI514-1247 VWRI514-1248 VWRI514-1252 VWRI514-1253 VWRI514-1254 VWRI514-1255 VWRI514-1256 VWRI514-1257 VWRI514-1258 VWRI514-1259 VWRI514-1260 VWRI514-1261 VWRI514-1262 VWRI514-1263 VWRI514-1264 VWRI514-1265 VWRI514-1266 VWRI514-1267 VWRI514-1268 VWRI514-1277 VWRI514-1269 VWRI514-1270 VWRI514-1271 VWRI514-1272 VWRI514-1273 VWRI514-1274 VWRI514-1275 VWRI514-1276 VWRI514-1278 VWRI514-1279 VWRI514-1280 VWRI514-1281
VWR®, Syringe Filters
过滤器 注射器过滤器
Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
  • Sterile blister packed filters are ideal for sterile filtration
  • Non sterile syringe filters are excellent for HPLC applications
  • Biosafe is according to Class VI plastics tests
  • Available with nylon, PES, PTFE, PVDF, CA, RC membrane, and GF (Glass Fibre)
  • Available with hydrophilic or hydrophobic PTFE
  • Each filter has a female Luer lock inlet and a male slip Luer outlet
Nylon and PTFE membranes are ideal for sample preparation and small volume chemicalMore Product Information