VWR®, Test Sieves, 305×60 mm, with Square Holes
供应商: VWR Collection
VWRI510-2929EA 6261 CNY
VWRI510-2929 VWRI510-2930 VWRI510-2931 VWRI510-2932 VWRI510-2933 VWRI510-2934 VWRI510-2935 VWRI510-2936 VWRI510-2937 VWRI510-2938 VWRI510-3300 VWRI510-3301 VWRI510-3302 VWRI510-3303 VWRI510-3304 VWRI510-3305 VWRI510-3306 VWRI510-3307 VWRI510-3308 VWRI510-3309 VWRI510-3310 VWRI510-3311 VWRI510-3312 VWRI510-3313 VWRI510-3314 VWRI510-3315 VWRI510-3316 VWRI510-3317 VWRI510-3318 VWRI510-3319 VWRI510-3320 VWRI510-3321 VWRI510-3322 VWRI510-3323 VWRI510-3324 VWRI510-3325 VWRI510-3326 VWRI510-3327 VWRI510-3328 VWRI510-3329 VWRI510-3330
VWR®, Test Sieves, 305×60 mm, with Square Holes
筛子 试验筛
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.