VWR®, Test Sieves, 305×60 mm
供应商: VWR Collection
VWRI510-2837EA 5766 CNY
VWRI510-2837 VWRI510-2838 VWRI510-2839 VWRI510-2840 VWRI510-2841 VWRI510-2842 VWRI510-2843 VWRI510-2844 VWRI510-2845 VWRI510-2846 VWRI510-2847 VWRI510-2848 VWRI510-2849 VWRI510-2850 VWRI510-2851 VWRI510-2852 VWRI510-2853 VWRI510-2854 VWRI510-2855 VWRI510-2856 VWRI510-2857 VWRI510-2858 VWRI510-2859 VWRI510-2860 VWRI510-2861 VWRI510-2862 VWRI510-2863 VWRI510-2864 VWRI510-2865 VWRI510-2866 VWRI510-2867 VWRI510-2868 VWRI510-2869 VWRI510-2870 VWRI510-2871 VWRI510-2872 VWRI510-2873 VWRI510-2874 VWRI510-2875 VWRI510-2876 VWRI510-2877 VWRI510-2878 VWRI510-2879 VWRI510-2880 VWRI510-2881 VWRI510-2882 VWRI510-2883 VWRI510-2884 VWRI510-2885 VWRI510-2886 VWRI510-2887 VWRI510-2888 VWRI510-2889 VWRI510-2890 VWRI510-2891 VWRI510-2892 VWRI510-2893 VWRI510-2894 VWRI510-2895 VWRI510-2896 VWRI510-2897 VWRI510-2898 VWRI510-2899 VWRI510-2900 VWRI510-2901 VWRI510-2902 VWRI510-2903 VWRI510-2904 VWRI510-2905 VWRI510-2906 VWRI510-2907 VWRI510-2908 VWRI510-2909 VWRI510-2910 VWRI510-2911 VWRI510-2912 VWRI510-2913 VWRI510-2914 VWRI510-2915 VWRI510-2916 VWRI510-2917 VWRI510-2918 VWRI510-2919 VWRI510-2920 VWRI510-2921 VWRI510-2922 VWRI510-2923 VWRI510-2924 VWRI510-2925 VWRI510-2926 VWRI510-2927 VWRI510-2928
VWR®, Test Sieves, 305×60 mm
筛子 试验筛
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.