VWR®, Test Sieves, 300×60 mm, with Square Holes
供应商: VWR Collection
VWRI510-2789EA 6261 CNY
VWRI510-2789 VWRI510-2790 VWRI510-2791 VWRI510-2792 VWRI510-2793 VWRI510-2794 VWRI510-2795 VWRI510-2796 VWRI510-2797 VWRI510-2798 VWRI510-2799 VWRI510-2801 VWRI510-2802 VWRI510-2803 VWRI510-2800 VWRI510-2804 VWRI510-2805 VWRI510-2806 VWRI510-2807 VWRI510-2808 VWRI510-2809 VWRI510-2810 VWRI510-2811 VWRI510-2812 VWRI510-2813 VWRI510-2814 VWRI510-2815 VWRI510-2816 VWRI510-2817 VWRI510-2818 VWRI510-2819 VWRI510-2820 VWRI510-2821 VWRI510-2822 VWRI510-2823 VWRI510-2824 VWRI510-2825 VWRI510-2826 VWRI510-2827 VWRI510-2828 VWRI510-2829
VWR®, Test Sieves, 300×60 mm, with Square Holes
筛子 试验筛
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.