VWR®, Test Sieves, 300×30 mm, with Square Holes
供应商: VWR Collection
VWRI510-2649EA 6261 CNY
VWRI510-2649 VWRI510-2650 VWRI510-2651 VWRI510-2652 VWRI510-2653 VWRI510-2654 VWRI510-2655 VWRI510-2656 VWRI510-2657 VWRI510-2658 VWRI510-2659 VWRI510-2660 VWRI510-2661 VWRI510-2662 VWRI510-2663 VWRI510-2664 VWRI510-2665 VWRI510-2666 VWRI510-2667 VWRI510-2668 VWRI510-2669 VWRI510-2670 VWRI510-2671 VWRI510-2672 VWRI510-2673 VWRI510-2674 VWRI510-2675 VWRI510-2676 VWRI510-2677 VWRI510-2678 VWRI510-2679 VWRI510-2680 VWRI510-2681 VWRI510-2682 VWRI510-2683 VWRI510-2684 VWRI510-2685 VWRI510-2686 VWRI510-2687 VWRI510-2688 VWRI510-2689
VWR®, Test Sieves, 300×30 mm, with Square Holes
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.