VWR®, Test Sieves, 450×70 mm
供应商: VWR Collection
VWRI510-1693EA 15286 CNY
VWRI510-1693 VWRI510-1687 VWRI510-1700 VWRI510-1699 VWRI510-1698 VWRI510-1697 VWRI510-1696 VWRI510-1701 VWRI510-1694 VWRI510-1704 VWRI510-1692 VWRI510-1691 VWRI510-1690 VWRI510-1689 VWRI510-1688 VWRI510-1695 VWRI510-1709 VWRI510-1716 VWRI510-1715 VWRI510-1714 VWRI510-1713 VWRI510-1712 VWRI510-1702 VWRI510-1710 VWRI510-1718 VWRI510-1708 VWRI510-1707 VWRI510-1706 VWRI510-1705 VWRI510-1685 VWRI510-1703 VWRI510-1711 VWRI510-1719 VWRI510-1686 VWRI510-1759 VWRI510-1766 VWRI510-1765 VWRI510-1764 VWRI510-1763 VWRI510-1762 VWRI510-1752 VWRI510-1760 VWRI510-1769 VWRI510-1758 VWRI510-1757 VWRI510-1756 VWRI510-1755 VWRI510-1754 VWRI510-1717 VWRI510-1761 VWRI510-1775 VWRI510-1781 VWRI510-1780 VWRI510-1779 VWRI510-1778 VWRI510-1767 VWRI510-1776 VWRI510-1768 VWRI510-1774 VWRI510-1773 VWRI510-1772 VWRI510-1771 VWRI510-1770 VWRI510-1751 VWRI510-1777 VWRI510-1726 VWRI510-1733 VWRI510-1732 VWRI510-1731 VWRI510-1730 VWRI510-1729 VWRI510-1753 VWRI510-1727 VWRI510-1736 VWRI510-1725 VWRI510-1724 VWRI510-1723 VWRI510-1722 VWRI510-1721 VWRI510-1720 VWRI510-1728 VWRI510-1742 VWRI510-1750 VWRI510-1749 VWRI510-1748 VWRI510-1747 VWRI510-1746 VWRI510-1745 VWRI510-1734 VWRI510-1743 VWRI510-1735 VWRI510-1741 VWRI510-1740 VWRI510-1739 VWRI510-1738 VWRI510-1737 VWRI510-1744
VWR®, Test Sieves, 450×70 mm
筛子 试验筛
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.