VWR®, Test Sieves, 203×32 mm
供应商: VWR Collection
VWRI510-0840EA 18174 CNY
VWRI510-0840 VWRI510-0841 VWRI510-0842 VWRI510-0843 VWRI510-0844 VWRI510-0845 VWRI510-0846 VWRI510-0847 VWRI510-0848 VWRI510-0849 VWRI510-0850 VWRI510-0851 VWRI510-0852 VWRI510-0853 VWRI510-0854 VWRI510-0855 VWRI510-0856 VWRI510-0857 VWRI510-0858 VWRI510-0859 VWRI510-0860 VWRI510-0861 VWRI510-0862 VWRI510-0863 VWRI510-0864 VWRI510-0865 VWRI510-0866 VWRI510-0867 VWRI510-0868 VWRI510-0869 VWRI510-0870 VWRI510-0871 VWRI510-0872 VWRI510-0873 VWRI510-0874 VWRI510-0875 VWRI510-0876 VWRI510-0877 VWRI510-0878 VWRI510-0879 VWRI510-0880 VWRI510-0881 VWRI510-0882 VWRI510-0883 VWRI510-0884 VWRI510-0885 VWRI510-0886 VWRI510-0887 VWRI510-0888 VWRI510-0889 VWRI510-0890 VWRI510-0891 VWRI510-0892 VWRI510-0893 VWRI510-0894 VWRI510-0895 VWRI510-0896 VWRI510-0897 VWRI510-0898 VWRI510-0899 VWRI510-0900 VWRI510-0901 VWRI510-0902 VWRI510-0903 VWRI510-0904 VWRI510-0905 VWRI510-0906 VWRI510-0907 VWRI510-0908 VWRI510-0909 VWRI510-0910
VWR®, Test Sieves, 203×32 mm
筛子 试验筛
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.