VWR®, Test Sieves, 100×45 mm
供应商: VWR Collection
VWRI510-0526EA 3412 CNY
VWRI510-0526 VWRI510-0527 VWRI510-0528 VWRI510-0529 VWRI510-0530 VWRI510-0531 VWRI510-0532 VWRI510-0533 VWRI510-0534 VWRI510-0535 VWRI510-0536 VWRI510-0537 VWRI510-0500 VWRI510-0501 VWRI510-0502 VWRI510-0503 VWRI510-0504 VWRI510-0505 VWRI510-0506 VWRI510-0507 VWRI510-0508 VWRI510-0509 VWRI510-0510 VWRI510-0511 VWRI510-0512 VWRI510-0513 VWRI510-0514 VWRI510-0515 VWRI510-0516 VWRI510-0517 VWRI510-0518 VWRI510-0519 VWRI510-0520 VWRI510-0521 VWRI510-0522 VWRI510-0523 VWRI510-0524 VWRI510-0525 VWRI510-0538 VWRI510-0539 VWRI510-0540 VWRI510-0541 VWRI510-0542 VWRI510-0543 VWRI510-0544 VWRI510-0545 VWRI510-0546 VWRI510-0547 VWRI510-0548 VWRI510-0549 VWRI510-0550 VWRI510-0551 VWRI510-0552 VWRI510-0553
VWR®, Test Sieves, 100×45 mm
筛子 试验筛
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.