VWR® VAPOUR-Line Vertical Autoclaves
供应商: VWR Collection
VWRI481-0689EA 0 CNY
VWRI481-0689 VWRI481-0690 VWRI481-0691 VWRI481-0692 VWRI481-0693 VWRI481-0694 VWRI481-0695 VWRI481-0696 VWRI481-0727 VWRI481-0728
VWR® VAPOUR-Line Vertical Autoclaves
干热灭菌器 高压灭菌器(蒸汽灭菌器)
VAPOUR- Line高压灭菌器适用于诸如开口或微密闭容器内的液体、仪器和固体、解包灭菌和废物灭菌。
  • Sterile filter for chamber venting is equipped as standard
  • Microprocessor controller with 10 pre-programmed protocols (5 pre-recorded freely modifiable and 5 protected by code)
  • TÜV-approved safety locking mechanism and thermal lock according to EN DIN 61010-2-43 with flexible sensor reference probe
  • Air removal by gravity (except M models)
  • Integrated exhaust steam condensation, water cooled and temperature controlled
  • Integrated memory chip for storing up to 150 cycles (later printout possible)