Stirring shafts
供应商: VWR Collection
VWRI441-0253EA 845 CNY
VWRI441-0253 VWRI441-0254 VWRI441-0255 VWRI441-0256 VWRI441-0257 VWRI441-0258 VWRI441-0259 VWRI441-0260 VWRI441-0261
Stirring shafts
搅拌器 Stirring Shafts
PTFE. For use with stirring blades. Chemically inert. The smooth surface reduces contamination and makes cleaning easier. It also prevents the agitation vessels from being scratched.
  • Excellent chemical resistance
  • Temperature resistant from −200 to 280 °C
订购须知 Stirring blades must be ordered separately.