VWR® Water Sampling Bottles, with Tamper Evident Cap
供应商: VWR Collection
VWRI331-0383EA 1785 CNY
VWRI331-0383 VWRI331-0384 VWRI331-0385 VWRI331-0386 VWRI331-0387 VWRI331-0388 VWRI331-0389 VWRI331-0390 VWRI331-0391 VWRI331-0392 VWRI331-0393 VWRI331-0394 VWRI331-0395 VWRI331-0396 VWRI331-0397 VWRI331-0398 VWRI331-0399 VWRI331-0400 VWRI331-0401 VWRI331-0402 VWRI331-0403 VWRI331-0404 VWRI331-0405 VWRI331-0406 VWRI331-0407 VWRI331-0408 VWRI331-0409
VWR® Water Sampling Bottles, with Tamper Evident Cap
水取样瓶
HDPE water sampling collection bottle.
  • Bottle made with HDPE resin
  • Natural bottle colour, with red cap
  • Supplied sterile
  • Sodium thiosulphate in powder form
  • Product labelled with unique barcode and sample collection write-on for traceability
  • Tamper-evident cap options with 33 mm pull type, 45 and 63 mm threaded cap
Wide neck design allows for easy sample collection. Product labelled with dosage and write-on details. These bottles are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the waterMore Product Information