VWR®, Wire basket with handle
供应商: VWR Collection
VWRI216-2254EA 2302 CNY
VWRI216-2254
VWR®, Wire basket with handle
篮筐 铁丝筐
This wire basket is made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Foldaway carrying handle with plastic hand grip
  • Robust design with two rods for base reinforcement
Suitable for transportation or sterilisation of laboratory goods.