VWR®, Workstation Dispenser, 3 Compartments
供应商: VWR Collection
VWRI216-2001EA 6589 CNY
VWRI216-2001
VWR®, Workstation Dispenser, 3 Compartments
箱子 分配器盒
Three-compartment dispenser, can be used to organise safety glasses, earplugs, soft covers (including bouffant caps, beard covers and shoe covers), or any small items that need to be kept within reach.
  • Three separate lift-up lids
  • Middle compartment has a reach-in hole for dispensing soft covers
  • Durable acrylic is see-through, so product levels are always visible
  • Set on counter or wall mount (screws included)
For labs, hospitals, doctor's offices, veterinarian offices, dentist offices, etc.