VWR®, PC Erlenmeyer Shaker Flasks
供应商: VWR Collection
VWRI214-0446EA 808 CNY
VWRI214-0446 VWRI214-0447 VWRI214-0448 VWRI214-0449 VWRI214-0450 VWRI214-0451 VWRI214-0452 VWRI214-0453 VWRI214-0476 VWRI214-0477 VWRI214-0478 VWRI214-0479 VWRI214-0480 VWRI214-0481
VWR®, PC Erlenmeyer Shaker Flasks
烧瓶 三角烧瓶
VWR® PC erlenmeyer shaker flasks provide excellent optical clarity and mechanical strength. They are ideal for shaker culture applications and storage.
  • Temperature range: –80 to +121 °C
  • Unlike glass, flasks will not break when dropped
  • Molded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Non-cytotoxic, DNase/RNase free and non-pyrogenic
  • Sterilized by irradiation SAL10-6 (ISO11137)
  • Shelf life: 3 years after month of production
Manufactured in a class 100,000 room environment underMore Product Information